SY3H3D67AD3D Synthonix Corporation

1-(2-Chloro-3-fluorophenyl)ethanamine hydrochloride

  • MDL number MFCD27923727
  • Cas Number : 1956306-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49235.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49235.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart