SY3H3D67AD3E Synthonix Corporation

1-(2-Chloro-6-fluorophenyl)ethanamine hydrochloride

  • MDL number MFCD21602735
  • Cas Number : 1375474-27-3
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49236.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49236.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart