SY3H3D67AD5F Synthonix Corporation

4-Cyanobenzamidine Hydrochloride

  • Cas Number : 117837-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49602.html

Safety Information

MSDS https://synthonix.com/49602.html
Availability details for
Quantity
  • Availability
  • Add to Cart