SY3H3D67AD63 Synthonix Corporation

2-Cyclohexylidenemalononitrile

  • MDL number MFCD00019512
  • Cas Number : 4354-73-8
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49712.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49712.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart