SY3H3D67AD64 Synthonix Corporation

(Z)-5H-Cyclopenta[b]pyridin-7(6H)-one O-methyl oxime

  • MDL number MFCD28975314
  • Cas Number : 1071727-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49731.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49731.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart