SY3H3D67AD65 Synthonix Corporation

2-(Cyclopropylamino)nicotinic acid

  • Cas Number : 639807-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49735.html

Safety Information

MSDS https://synthonix.com/49735.html
Availability details for
Quantity
  • Availability
  • Add to Cart