SY3H3D67AD67 Synthonix Corporation

2-Chloro-4-morpholinopyrimidine-5-carbaldehyde

  • Cas Number : 1820740-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49758.html

Safety Information

MSDS https://synthonix.com/49758.html
Availability details for
Quantity
  • Availability
  • Add to Cart