SY3H3D67AD6D Synthonix Corporation

(R)-5-Chloro-2,3-dihydro-1H-inden-1-amine

  • Cas Number : 812695-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49774.html

Safety Information

MSDS https://synthonix.com/49774.html
Availability details for
Quantity
  • Availability
  • Add to Cart