SY3H3D67AD70 Synthonix Corporation

4-Chloro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD08544170
  • Cas Number : 68755-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49777.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49777.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart