SY3H3D67AD71 Synthonix Corporation

6-Chloro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD08544166
  • Cas Number : 1191908-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49778.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49778.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart