SY3H3D67AD7F Synthonix Corporation

1-(2-Chloroethyl)-3-methylbenzene

  • MDL number MFCD11110036
  • Cas Number : 39199-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49969.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49969.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart