SY3H3D67AD84 Synthonix Corporation

(4-Chloro-3-(dimethylcarbamoyl)phenyl)boronic acid

  • MDL number MFCD07783868
  • Cas Number : 871332-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Light-brown to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50103.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50103.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart