SY3H3D67AD9A Synthonix Corporation

(5-Chloro-2-propoxyphenyl)boronic acid

  • MDL number MFCD06411348
  • Cas Number : 849062-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50294.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50294.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart