SY3H3D67AD9E Synthonix Corporation

4-Cyano-4-(((ethylthio)carbonothioyl)thio)pentanoic acid

  • Cas Number : 1137725-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50373.html

Safety Information

MSDS https://synthonix.com/50373.html
Availability details for
Quantity
  • Availability
  • Add to Cart