SY3H3D67AD9F Synthonix Corporation

1-Cyclohexyl-1H-imidazole

  • MDL number No MDL found
  • Cas Number : 67768-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50394.html

Safety Information

MSDS https://synthonix.com/50394.html
Availability details for
Quantity
  • Availability
  • Add to Cart