SY3H3D67ADA1 Synthonix Corporation

6-((3-Chloropropyl)amino)-1,3-dimethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD00453355
  • Cas Number : 34654-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50422.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50422.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart