SY3H3D67ADA7 Synthonix Corporation

2-Cyclohexylpropanoic acid

  • MDL number MFCD00268172
  • Cas Number : 6051-13-4
  • Molecular Weight : 156.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50503.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50503.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart