SY3H3D67ADA8 Synthonix Corporation

9-Chloronon-1-ene

  • Cas Number : 872-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50527.html

Safety Information

MSDS https://synthonix.com/50527.html
Availability details for
Quantity
  • Availability
  • Add to Cart