SY3H3D67ADAA Synthonix Corporation

Cyclohexyltrimethoxysilane

  • MDL number MFCD01074466
  • Cas Number : 17865-54-2
  • Molecular Weight : 204.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50616.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50616.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart