SY3H3D67ADB5 Synthonix Corporation

Cyclopenta[2,1-b:3,4-b']dithiophen-4-one

  • MDL number MFCD16619150
  • Cas Number : 25796-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Yellow red - Deep yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50662.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50662.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart