SY3H3D67ADC3 Synthonix Corporation

2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile

  • MDL number MFCD04115850
  • Cas Number : 886496-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50739.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50739.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart