SY3H3D67ADC8 Synthonix Corporation

1-Chloro-5-iodoisoquinoline

  • Cas Number : 1379322-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50744.html

Safety Information

MSDS https://synthonix.com/50744.html
Availability details for
Quantity
  • Availability
  • Add to Cart