SY3H3D67ADD9 Synthonix Corporation

5-Chloro-7-methylindoline-2,3-dione

  • MDL number MFCD00047218
  • Cas Number : 14389-06-1
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-7-methyl-1H-indole-2,3-dione
Inchi Code 1S/C9H6ClNO2/c1-4-2-5(10)3-6-7(4)11-9(13)8(6)12/h2-3H,1H3,(H,11,12,13)
Inchi Key LDFQLYHDZZPAGN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Yellow to red to deep-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/50876.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/50876.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart