SY3H3D67AE03 Synthonix Corporation

5-Chloro-7-fluoro-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18647727
  • Cas Number : 1273613-81-2
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51027.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51027.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart