SY3H3D67AE04 Synthonix Corporation

6-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one

  • Cas Number : 174603-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51028.html

Safety Information

MSDS https://synthonix.com/51028.html
Availability details for
Quantity
  • Availability
  • Add to Cart