SY3H3D67AE09 Synthonix Corporation

2-Chloroquinolin-4-amine

  • MDL number MFCD08705643
  • Cas Number : 80947-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51069.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51069.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart