SY3H3D67AE2F Synthonix Corporation

4-Cyano-3-methylbenzoic acid

  • Cas Number : 73831-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51177.html

Safety Information

MSDS https://synthonix.com/51177.html
Availability details for
Quantity
  • Availability
  • Add to Cart