SY3H3D67AE30 Synthonix Corporation

(1-Chloroisoquinolin-4-yl)boronic acid

  • Cas Number : 848841-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51188.html

Safety Information

MSDS https://synthonix.com/51188.html
Availability details for
Quantity
  • Availability
  • Add to Cart