SY3H3D67AE3E Synthonix Corporation

(E)-3-Chlorocinnamaldehyde

  • MDL number MFCD07437882
  • Cas Number : 56578-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51317.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51317.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart