SY3H3D67AE43 Synthonix Corporation

2-Cyano-2-phenylacetamide

  • MDL number MFCD17240132
  • Cas Number : 771-84-6
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51349.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51349.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart