SY3H3D67AE54 Synthonix Corporation

7-Chloro-3,4-dihydroquinolin-2(1H)-one

  • Cas Number : 14548-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51528.html

Safety Information

MSDS https://synthonix.com/51528.html
Availability details for
Quantity
  • Availability
  • Add to Cart