SY3H3D67AE59 Synthonix Corporation

Isochroman-1-one

  • MDL number MFCD00799232
  • Cas Number : 4702-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51549.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51549.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart