SY3H3D67AE5C Synthonix Corporation

3-(5-Chloro-2-fluorophenyl)propanoic acid

  • MDL number MFCD18393096
  • Cas Number : 135832-52-9
  • Molecular Weight : 202.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(5-chloro-2-fluorophenyl)propanoic acid
Inchi Code 1S/C9H8ClFO2/c10-7-2-3-8(11)6(5-7)1-4-9(12)13/h2-3,5H,1,4H2,(H,12,13)
Inchi Key WJGBGNPQWOGIER-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/51607.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/51607.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart