SY3H3D67AE66 Synthonix Corporation

3-Chloro-4-methoxy-1-methyl-1H-indazole

  • MDL number MFCD27923528
  • Cas Number : 1780480-20-7
  • Molecular Weight : 196.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51696.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51696.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart