SY3H3D67AE6B Synthonix Corporation

N-(5-Cyano-2-methylpyridin-3-yl)acetamide

  • MDL number MFCD28134464
  • Cas Number : 1628557-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51721.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51721.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart