SY3H3D67AE6C Synthonix Corporation

4-[6-chloro-2-(trifluoromethyl)pyrimidin-4-yl]morpholine

  • Cas Number : 1189444-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51737.html

Safety Information

MSDS https://synthonix.com/51737.html
Availability details for
Quantity
  • Availability
  • Add to Cart