SY3H3D67AE75 Synthonix Corporation

1-(2-Chloro-4-methoxyphenyl)ethanone

  • Cas Number : 41068-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51927.html

Safety Information

MSDS https://synthonix.com/51927.html
Availability details for
Quantity
  • Availability
  • Add to Cart