SY3H3D67AE78 Synthonix Corporation

2-(2-(Chloromethyl)phenyl)acetic acid

  • Cas Number : 95335-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51930.html

Safety Information

MSDS https://synthonix.com/51930.html
Availability details for
Quantity
  • Availability
  • Add to Cart