SY3H3D67AE81 Synthonix Corporation

1-(2-Chlorophenyl)propan-2-one

  • MDL number MFCD00045175
  • Cas Number : 6305-95-9
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chlorophenyl)acetone
Inchi Code 1S/C9H9ClO/c1-7(11)6-8-4-2-3-5-9(8)10/h2-5H,6H2,1H3
Inchi Key LWGNDIMNCPMZOF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/52039.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/52039.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart