SY3H3D67AE93 Synthonix Corporation

1-(3-Chlorobenzo[b]thiophen-2-yl)-N-methylmethanamine hydrochloride

  • MDL number MFCD13193901
  • Cas Number : 132740-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52360.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52360.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart