SY3H3D67AE96 Synthonix Corporation

4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-7-yl acetate

  • MDL number MFCD11112163
  • Cas Number : 945666-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52370.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52370.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart