SY3H3D67AE98 Synthonix Corporation

4-(4-Chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD00854139
  • Cas Number : 22518-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52398.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52398.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart