SY3H3D67AE9A Synthonix Corporation

Chroman-6-carbaldehyde

  • MDL number MFCD08146589
  • Cas Number : 55745-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52424.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52424.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart