SY3H3D67AEA5 Synthonix Corporation

3-(4-Chlorophenyl)piperazin-2-one

  • MDL number MFCD07373585
  • Cas Number : 86147-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52520.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52520.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart