SY3H3D67AEAA Synthonix Corporation

4-Chloro-1-(4-hydroxyphenyl)butan-1-one

  • MDL number MFCD00002362
  • Cas Number : 7150-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52637.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52637.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart