SY3H3D67AEAD Synthonix Corporation

3-Chloro-4-isopropoxybenzoic acid

  • Cas Number : 213598-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52660.html

Safety Information

MSDS https://synthonix.com/52660.html
Availability details for
Quantity
  • Availability
  • Add to Cart