SY3H3D67AEB3 Synthonix Corporation

6-Chloro-1,2,3,4-tetrahydronaphthalen-1-amine

  • Cas Number : 1337048-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52743.html

Safety Information

MSDS https://synthonix.com/52743.html
Availability details for
Quantity
  • Availability
  • Add to Cart