SY3H3D67AEBB Synthonix Corporation

Cyclohexane-1,2,4,5-tetracarboxylic acid

  • MDL number MFCD00435556
  • Cas Number : 15383-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52864.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52864.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart