SY3H3D67AF46 Synthonix Corporation

4-Chloro-7-cyclopentyl-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD09909354
  • Cas Number : 213745-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54391.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54391.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart