SY3H3D67AF4A Synthonix Corporation

3-(4-Chlorophenyl)-1,4-dioxane-2-carboxylic acid

  • Cas Number : 1706443-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54471.html

Safety Information

MSDS https://synthonix.com/54471.html
Availability details for
Quantity
  • Availability
  • Add to Cart